missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nutrient Broth with Glucose (Tube)

Cultivate non fastidious micro-organisms for testing food, milk and dairy products using these ready prepared tubes.

Brand:  Thermo Scientific™ TV5003D

47.43 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 10717173

  • 48.65€ / Pack of 50
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Cultivate non-fastidious microorganisms for testing food, milk and dairy products with Thermo Scientific™ Oxoid™ Nutrient Broth with Glucose ready prepared tubes. This non-selective, fluid medium contains beef extracts combined with peptone, glucose, yeast extracts and sodium chloride to provide the necessary nitrogen compounds, carbon, vitamins and some trace ingredients necessary for the growth of bacteria.

Remove the need for autoclaving and dispensing with convenient, pre-filled and ready prepared tubes of Nutrient Broth.

  • Suitable for general cultivation of less fasidious microorganisms.
  • Convenient: the pre-filled tubes are ready for immediate use.
  • Simple: easy-to-use with minimum training.
  • Long shelf life: up to 12 months.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Signal yellow, transparent
Nutrient Broth with Glucose (Tube)
Germany
50 tubes in a box
2 to 25°C
Poured Tube
Microbiology Media
50/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number6251791 Certificate TypeCertificate of Analysis Date31/03/2025 Catalog NumberTV5003D
Lot Number6248836 Certificate TypeCertificate of Analysis Date24/03/2025 Catalog NumberTV5003D
Lot Number6246039 Certificate TypeCertificate of Analysis Date17/03/2025 Catalog NumberTV5003D
Lot Number6241723 Certificate TypeCertificate of Analysis Date10/03/2025 Catalog NumberTV5003D
Lot Number6239004 Certificate TypeCertificate of Analysis Date03/03/2025 Catalog NumberTV5003D
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Nutrient Broth with Glucose (Tube) >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.