Learn More
Thermo Scientific™ 96 Black Plate, Non-Treated Coverglass Surface, No Lid, Non-Sterile, Pack of 5
Color:Black, Clear Bottom; Sterility; Non-sterile; Lid, Without Lid; Type: Optical Coverglass Base
Brand: Thermo Scientific™ 265300
2640.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Achieve optimal fluorescence and luminescence in cell culture and microscopic applications with the Thermo Scientific™ Nunc™ MicroWell™ 96-Well Optical-Bottom Plates with Coverglass Base, featuring a polystyrene black or white upper structure with a polystyrene film at the bottom of the plate.
Features:
- Background, autoflorescence and crosstalk are improved
- Black microplates for florescence measurements, with minimum back-scattered light and background fluorescence
- White microplates for luminescence measurements, with maximum reflection and minimal autoluminescence
- Cell culture surface is ideal for cell-based assays
- CC2 surface modification closely mimics Poly-D-Lysine and is superior for attachment and growth of fastidious cells
- Customized bar coding available
Specifications
Custom Barcoding Available | |
Without Lid | |
5/Pk., 30/Cs. | |
Black | |
Polystyrene | |
Non-sterile | |
Flat Bottom (F-well) | |
5 |
All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) | |
8 x 12 | |
Nunc | |
Optical Coverglass Base | |
96 | |
Non-treated | |
30 | |
Case of 30 |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1411725 | Certificate of Compliance | 23/04/2025 | 265300 |
1312961 | Certificate of Compliance | 11/12/2024 | 265300 |
1399413 | Certificate of Compliance | 02/10/2024 | 265300 |
1391909 | Certificate of Compliance | 30/09/2023 | 265300 |
1350584 | Certificate of Compliance | 17/08/2022 | 265300 |