missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific™ Nunc™ MicroWell™ 96-Well, Nunclon Delta-Treated, Flat-Bottom Microplate Product Code.: 10525411
Add to basket

Thermo Scientific™ Nunc™ MicroWell™ 96-Well, Nunclon Delta-Treated, Flat-Bottom Microplate
BIOPHARMA

Product Code. 10525411
160/Cs.
Click to view available options
Quantity:
160/Cs.
50/Cs.
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price

Product Code. 10525411

Brand: Thermo Scientific™ 137103

Add to basket
Add to basket
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price

Black, sterile microplates with lid. Ideal for fluorescence measurements, with minimum back-scattered light and background fluorescence.

Ensure consistent and reliable results using the high quality Thermo Scientific™ Nunc™ F96 MicroWell™ Black and White Polystyrene Plate. Black and white plates are optimized for robotics, plate readers and liquid handling systems and designed for key imaging characteristics. Available with or without cell culture treatment.

  • Background, autoflorescence and crosstalk are improved when you use Nunc Black or White microplates
  • Black microplates are recommended for florescence measurements; with minimum back-scattered light and background fluorescence
  • White microplates are recommended for luminescence measurements; with maximum reflection and minimal autoluminescence
  • Working volume range: 50 to 250μL/well
  • Choice of Thermo Scientific™ Nunclon™ Delta surface for cell-based assays
  • Customized bar coding is available
TRUSTED_SUSTAINABILITY

Specifications

Barcode Custom Barcoding Available
Certifications/Compliance All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
Certified Nunclon Delta certified for monolayer formation, cloning efficiency, non-cytotoxic, non-pyrogenic, and sterility
Lid With Lid
Array 8 x 12
Packaging 10 Pk., 160 Cases
Product Line Nunc
Color Black
Product Type Microplate
Material Polystyrene
Volume (Metric) Well 400 μL
No. of Wells 96
Volume (Metric) Working 50 to 250 μL/well
Sterility Sterile
Surface Treatment Nunclon Delta
Well Shape Flat Bottom (F-well)
No. per Case 160
No. per Pack 10
Quantity 160/Cs.
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number184328 Certificate TypeCertificate of Compliance Date30/01/2025 Catalog Number137103
Lot Number171385 Certificate TypeCertificate of Compliance Date02/12/2024 Catalog Number137103
Lot Number179433 Certificate TypeCertificate of Compliance Date02/12/2024 Catalog Number137103
Lot Number175322 Certificate TypeCertificate of Compliance Date02/12/2024 Catalog Number137103
Lot Number184328 Certificate TypeCertificate of Irradiation Date15/01/2025 Catalog Number137103
  • Multiple Manufacturers
    Fall in love with big savings
    Promo Code: 24107

    Expires2025-03-28

    Days

    14

    Hours

    1

    Minutes

    52

    Seconds

    55
    New Offer
    Shop Now
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Nunc™ MicroWell™ 96-Well, Nunclon Delta-Treated, Flat-Bottom Microplate >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.