missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ gDNA Clean-up, Mini kit for DNA clean up and concentration

Concentrate DNA samples more easily and quickly, compared to microdialysis filtrations units. Macherey-Nagel™ NucleoSpin™ gDNA Clean-up Kit cleans and concentrates large-sized genomic DNA from pre-purified DNA from organic extraction and from enzymatic extractions.

Brand:  Macherey-Nagel™ 740230.50


 View more versions of this product

Product Code. 12783391

  • 138.00€ / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The NucleoSpin™ gDNA Clean-up kit is designed for the rapid purification of previously isolated small and especially high molecular weight genomic DNA. It is used to clean-up and concentrate genomic DNA after crude extraction methods, for example using Trizol, or after enzymatic, or chemical reactions. The time-saving procedures enables a preparation time of just 15 minutes for 10 preps.

  • No need for organic denaturants or chloroform extractions
  • Any impurities like phenol, enzymes, salts, dyes, labels, nucleotides, small oligonucleotides, and even up to 5% detergents (e.g., SDS, Triton, Tween, Lauroylsarcosin) are removed completely
  • Binding Buffer DB and Wash Buffer DW are developed to allow a very gentle binding and washing
  • High recovery rates up to 90% for high molecular weight DNA (up to 50 kbp) as well as for small DNA fragments down to 100 bp
  • The eluted DNA is ready to use for downstream applications such as PCR, endonuclease restriction, Southern Blotting and labeling
Kit components:
  • NucleoSpin™ gDNA Clean-up Columns, Collection Tubes (2 mL), Binding Buffer DB, Wash Buffer DW, Elution Buffer DE
Specifications

Specifications

Mini Spin Column
DNA (Genomic)
Elution volume: 50–100μL
NucleoSpin™ gDNA Clean-up Columns (50), 2mL Collection Tubes (50), Binding Buffer DB (25mL), Wash Buffer DW Concentrate (25mL), Elution Buffer DE (13mL)
Nucleo Spin gDNA Clean Up
<150 μL solution containing <25 μg DNA
Silica Membrane Technology
Recovery rate: 80–90%
Nucleo Spin gDNA Clean Up
50
A260/A280: 1.8–1.9
<15 min/10 preps
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2

Hazard Statement
  • H225-Highly flammable liquid and vapour.

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Macherey-Nagel™ NucleoSpin™ gDNA Clean-up, Mini kit for DNA clean up and concentration > No. of Reactions: 50

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.