missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ NS 9283

Positive allosteric modulator of α4β2 nAChRs

269.00€ - 1060.00€

Specifications

Purity >99%
Product Type NS 9283
Molecular Weight (g/mol) 248.24
Inhibitors NS 9283
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18756248
View Documents
Tocris Bioscience™
5112/10
10 mg
269.00€
10mg
Please to purchase this item. Need a web account? Register with us today!
18765999
View Documents
Tocris Bioscience™
5112/50
50 mg
1060.00€
50mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Positive allosteric modulator of α4β2 receptor; increases the potency of Ach-evoked currents ∽60 fold without effecting the maximum efficacy (HEK293-hα4β2 cells). Reduces the rate of recovery from desensitization and slows the rate of deactivation. Displays a synergistic effect on the Emax when given with NS206 at 3α:2β receptors.
Specifications

Specifications

>99%
NS 9283
248.24
NS 9283
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.