missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ WDR5 Protein
Highly purified. Generating reliable and reproducible results.
394.00€ - 1025.00€
Specifications
Concentration | 1mg/mL |
---|---|
For Use With (Application) | SDS-PAGE |
Formulation | Liquid. In 20mM Tris-HCl buffer (pH 8.0) containing 10% glycerol, 2mM DTT, 0.1M NaCl. |
Molecular Weight (g/mol) | 38.8kDa |
Purification Method | SDS-PAGE |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18255354
|
Novus Biologicals™
NBP1-50848-0.1MG |
0.1 mg |
394.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18202083
|
Novus Biologicals™
NBP1-50848-0.5MG |
0.5 mg |
1025.00€
0.50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-334 of Human WDR5 The Recombinant Human WDR5 Protein is derived from E. coli. The Recombinant Human WDR5 Protein has been validated for the following applications: SDS-Page.Specifications
1mg/mL | |
Liquid. In 20mM Tris-HCl buffer (pH 8.0) containing 10% glycerol, 2mM DTT, 0.1M NaCl. | |
SDS-PAGE | |
RUO | |
Unconjugated |
SDS-PAGE | |
38.8kDa | |
Store at -80°C. Avoid freeze-thaw cycles. | |
WDR5 | |
>95% |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title