missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ WDR5 Protein
Highly purified. Generating reliable and reproducible results.
Brand: Novus Biologicals™ NBP1-50848-0.5MG
This item is not returnable.
View return policy
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-334 of Human WDR5 The Recombinant Human WDR5 Protein is derived from E. coli. The Recombinant Human WDR5 Protein has been validated for the following applications: SDS-Page.Specifications
1mg/mL | |
Liquid. In 20mM Tris-HCl buffer (pH 8.0) containing 10% glycerol, 2mM DTT, 0.1M NaCl. | |
SDS-PAGE | |
Store at -80°C. Avoid freeze-thaw cycles. | |
WDR5 | |
>95% |
SDS-PAGE | |
38.8kDa | |
0.5 mg | |
RUO | |
Unconjugated |