missing translation for 'onlineSavingsMsg'
Learn More

Nitrite Standard Solution, Chem Lab™

Product Code. 11989102
Click to view available options
Quantity:
100 mL
500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Nitrite standard solution 1000 µg/ml for IC (Ion HIQU)

Chemical Identifiers

CAS 7632-00-0
Molecular Formula NNaO2
Molecular Weight (g/mol) 69.00
InChI Key LPXPTNMVRIOKMN-UHFFFAOYSA-M
IUPAC Name sodium nitrite
SMILES [Na+].[O-]N=O

Specifications

Concentration Solution contains 1000 µg/ml of NO2-
Quantity 500 mL
Color Colorless
Traceability to NIST Reference standards acc. NIST SRM 136f
Odor Odorless
pH ±9
Boiling Point 100 C
Solubility Soluble in water
Physical Form Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.