Learn More
Tocris Bioscience™ NITD008
Antiviral; selective flavivirus inhibitor
Brand: Tocris Bioscience™ 6045/1
Description
NITD 008 is a selective flavivirus inhibitor. Adenosine analog. Inhibits dengue virus (EC50 = 0.64 μM), hepatitis C virus and cellular entry of Zika virus. Reduces increases of cytokines and NS1 and prevents death in dengue-infected mice. Active in vivo. Orally bioavailable.Specifications
7-(2-C-Ethynyl-β-D-ribofuranosyl)-7 H-pyrrolo[2,3-d]pyrimidin-4-amine | |
1044589-82-3 | |
>98% | |
1 mg |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.