Learn More
Gibco™ NaOH, 1.0 M
Gibco NaOH solutions offer more options for supply security and dual sourcing of the highest volume process liquid in bioprocessing manufacturing.
Brand: Gibco™ A4782901
480.70 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
Gibco NaOH solutions offer more options for supply security and dual sourcing of the highest volume process liquid in bioprocessing manufacturing. Available in three different grades and two off-the-shelf sizes, these solutions provide outsourcing efficiencies to help reclaim time, space, and other core resources.
Features of Gibco NaOH solutions include:
• 0.1 M, 0.5 M, and 1 M concentrations
• Multi-compendial grade raw materials
• cGMP manufacturing to 21CFR820
• 20 L and 200 L ready-to-use single-use bioprocessing containers
Flexibility
Gibco NaOH solutions provide outsourcing efficiencies for customers manufacturing internally but seeking to partially outsource, as well as for customers with batch sizes that do not justify custom ordering and customers already outsourcing to Thermo Fisher Scientific but seeking additional flexibility. They are produced in the most commonly used concentrations and single-use containers.
Applications of Gibco NaOH solutions
• pH adjustment
• Facility/equipment sanitization
• Chromatography column cleaning, sanitization and storage

Specifications
Sodium Hydroxide | |
0°C | |
Clear | |
≥12 to ≤14 | |
HNaO | |
Caustic soda, Acidity regulatory, Lye or Soda lye | |
[OH-].[Na+] | |
40.00 | |
Compendial |
1310-73-2 | |
100°C | |
1.0 M | |
20 L | |
Bioprocess Container | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
sodium hydroxide | |
95 to 100% |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2544650 | Certificate of Analysis | 29/01/2025 | A4782902,A4782901 |
2534582 | Certificate of Analysis | 29/01/2025 | A4782902,A4782901 |
2550674 | Certificate of Analysis | 29/01/2025 | A4782902,A4782901 |
2440148 | Certificate of Analysis | 29/01/2025 | A4782902,A4782901 |
2529905 | Certificate of Analysis | 29/01/2025 | A4782902,A4782901 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.