Learn More
Gibco™ NaOH, 0.1 M
Gibco NaOH solutions offer more options for supply security and dual sourcing of the highest volume process liquid in bioprocessing manufacturing.
484.00€ - 1445.00€
Specifications
Chemical Name or Material | Sodium Hydroxide |
---|---|
CAS | 1310-73-2 |
Melting Point | 0°C |
Boiling Point | 100°C |
Color | Clear |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17850925
|
Gibco™
A4782601 |
20 L |
484.00€
20 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17870925
|
Gibco™
A4782602 |
200 L |
1445.00€
200 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Gibco NaOH solutions offer more options for supply security and dual sourcing of the highest volume process liquid in bioprocessing manufacturing. Available in three different grades and two off-the-shelf sizes, these solutions provide outsourcing efficiencies to help reclaim time, space, and other core resources.
Features of Gibco NaOH solutions include:
• 0.1 M, 0.5 M, and 1 M concentrations
• Multi-compendial grade raw materials
• cGMP manufacturing to 21CFR820
• 20 L and 200 L ready-to-use single-use bioprocessing containers
Flexibility
Gibco NaOH solutions provide outsourcing efficiencies for customers manufacturing internally but seeking to partially outsource, as well as for customers with batch sizes that do not justify custom ordering and customers already outsourcing to Thermo Fisher Scientific but seeking additional flexibility. They are produced in the most commonly used concentrations and single-use containers.
Applications of Gibco NaOH solutions
• pH adjustment
• Facility/equipment sanitization
• Chromatography column cleaning, sanitization and storage
Specifications
Sodium Hydroxide | |
0°C | |
Clear | |
≥12 to ≤14 | |
Bioprocess Container | |
95 to 100% |
1310-73-2 | |
100°C | |
0.1 M | |
NaOH | |
Caustic soda, Acidity regulatory, Lye or Soda lye | |
Compendial |
Safety and Handling
missing translation for 'shelfLife' : 12 months
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.