Learn More
Thermo Scientific NanoDrop™ Reagents
Certified standards for verifying instrument performance to factory specifications are available separately.
Brand: Thermo Scientific CHEM-PV-1
Description
Certified standards for verifying instrument performance to factory specifications are available separately.
Features include:- Solution with certified absorbance values
- Use to create a record of correct photometric performance of a NanoDrop One spectrophotometer
Specifications
NanoDrop PV-1 Performance Verification Solution | |
NanoDrop Ultra, NanoDrop One and NanoDrop OneC spectrophotometers | |
Store at room temperature |
Product Suggestions
Customers who viewed this item also viewed