Learn More
Thermo Scientific™ Nalgene™ Locator™ Plus Racks for 81-Ampule Cryogenic Box
Take advantage of efficient, high-volume storage with Thermo Scientific™ Nalgene™ Locator™ Plus Racks for 81-Ampule Cryogenic Box.
Brand:  Thermo Scientific™ HR509X20A-70
266.90 EUR valid until 2025-04-30
Use promo code "24119" to get your promotional price.
 View more versions of this product
Product Code. 12787065
  • 311.55€ / Each
Estimated Shipment: 09-07-2025
to see stock.
Explore more special offers
Description
Take advantage of efficient, high-volume storage with Thermo Scientific™ Nalgene™ Locator™ Plus Racks for 81-Ampule Cryogenic Box.
  • Designed for long-term storage
  • Features an indexed frame and box organizational system
  • Unique hanging rack promotes easy sample retrieval
  • Suited for both manual and computerized inventory record-keeping methods
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Gray
5.25 in.
Cryogenic Box, 81-Ampule
5.25 in.
For: Locator 4 Plus
Locator™ Plus Cryogenic Rack
Documents
One moment while we fetch your results.
Product Title
Thermo Scientific™ Nalgene™ Locator™ Plus Racks for 81-Ampule Cryogenic Box > L x W: 5.25 x 5.25 ; Thermolyne No.: HR509X20A 70; For: Locator 4 Plus