Learn More
Thermo Scientific™ Nalgene™ BioTransport Carrier Handles
Securely hold a fully loaded carrier with Thermo Scientific™ Nalgene™ BioTransport Carrier Handles, which fold conveniently to the side for storage.
Brand: Thermo Scientific™ 7136-0001
94.74 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
Securely hold a fully loaded carrier with Thermo Scientific™ Nalgene™ BioTransport Carrier Handles, which fold conveniently to the side for storage. These handles fit the BioTransport carrier for easy one-handed carrying.
- Simple to put on and take off
- Autoclavable stainless steel
- Includes: Handle only. Biotransport Carrier sold separately, Cat. No. 7135-0001
Please recycle the corrugate cartons and any LDPE bags your Nalgene products are delivered in.
Specifications
BioTransport Carrier Handle | |
Bio Transport Carrier Handle | |
Autoclavable |
Promotions
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85%