Learn More
Macherey-Nagel™ N9 Polypropylene Blue Screw Caps
Seal vials with screw caps with a secure septum to receive needles. The Macherey-NagelTM N9 pollypropylene blue screw caps come with a choice of septa.
Brand:  Macherey-Nagel™ 702287.1
 View more versions of this product
Product Code. 10282660
  • 25.60€ / Pack of 100
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Screw closures N 9
  • Large range of screw closures with different septum materials and cap colors
  • Bonded screw caps (septa firmly connected with the cap; cannot be removed) – advantage: thick (blunt) HPLC needles cannot push the septum into the vial
  • Polymagnetic screw closure 702166 for CTC GC PAL
Specifications
Specifications
Blue
No
1 mm
Polypropylene, PTFE, Silicone
White Silicone/Red PTFE
N9
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Macherey-Nagel™ N9 Polypropylene Blue Screw Caps > Septum: Silicone white/PTFE red