missing translation for 'onlineSavingsMsg'
Learn More
N-Isopropylhydroxylamine hydrochloride, 97%, Thermo Scientific Chemicals Product Code.: 10287450

N-Isopropylhydroxylamine hydrochloride, 97%, Thermo Scientific Chemicals

Product Code. 10287450
5 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10287450

Brand: Thermo Scientific Acros 276260050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50632-53-6
Molecular Formula C3H9NO·HCl
Molecular Weight (g/mol) 111.57
MDL Number MFCD00012599
InChI Key BYXUIKZQGOPKFR-UHFFFAOYSA-N
Synonym n-isopropylhydroxylamine hydrochloride, n-isopropyl hydroxylamine, n-isopropylhydroxylamine hcl, 2-propanamine, n-hydroxy-, hydrochloride, ccris 4317, i-propylhydroxylamine hydrochloride, isopropylhydroxylamine hydrochloride, n-hydroxypropan-2-amine hydrochloride, n-isopropyl hydroxylamine hydrochloride, n-isopropyl-hydroxylamine hydrochloride
PubChem CID 170873
IUPAC Name N-propan-2-ylhydroxylamine;hydrochloride
SMILES CC(C)NO.Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 68°C to 72°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 96% min. (Argentometry)
Packaging Glass bottle
Linear Formula (CH3)2CHNHOH·HCl
Quantity 5 g
Beilstein 04, 538
Formula Weight 111.57
Percent Purity 97%
Physical Form Adhering Crystalline Powder
Chemical Name or Material N-Isopropylhydroxylamine hydrochloride, 97%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0430590 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number276260050,276260010
Lot NumberA0403379 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number276260050,276260010
Lot NumberA0387340 Certificate TypeCertificate of Analysis Date18/08/2021 Catalog Number276260050,276260010
Lot NumberA0427456 Certificate TypeCertificate of Analysis Date08/03/2021 Catalog Number276260050,276260010
Lot NumberA0425572 Certificate TypeCertificate of Analysis Date23/12/2020 Catalog Number276260050,276260010
compliance-icons
Product Identifier
  • N-Isopropylhydroxylamine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-Isopropylhydroxylamine hydrochloride, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.