missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Fmoc-D-aspartic acid 4-tert-butyl ester, 98%, Thermo Scientific Chemicals
26.10€ - 286.00€
Chemical Identifiers
CAS | 112883-39-3 |
---|---|
Molecular Formula | C23H25NO6 |
Molecular Weight (g/mol) | 411.45 |
MDL Number | MFCD00077050 |
InChI Key | FODJWPHPWBKDON-UHFFFAOYNA-N |
Synonym | fmoc-d-asp otbu-oh, r-2-9h-fluoren-9-yl methoxy carbonyl amino-4-tert-butoxy-4-oxobutanoic acid, fmoc-d-asp obut-oh, fmoc-d-aspartic acid beta-tert-butyl ester, n-alpha-fmoc-d-aspartic acid beta-t-butyl ester, 2r-4-tert-butoxy-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-oxobutanoic acid, fmoc-dasp otbu, ambotzfaa1309, fmoc-dasp otbu-oh, pubchem10009 |
PubChem CID | 7018816 |
SMILES | CC(C)(C)OC(=O)CC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15487316
|
Thermo Scientific Alfa Aesar
H62356.MD |
250 mg | N/A | ||||||
15408888
|
Thermo Scientific Alfa Aesar
H62356.03 |
1 g |
74.30€
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15418888
|
Thermo Scientific Alfa Aesar
H62356.06 |
5 g |
286.00€
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
112883-39-3 | |
411.45 | |
FODJWPHPWBKDON-UHFFFAOYNA-N | |
7018816 |
C23H25NO6 | |
MFCD00077050 | |
fmoc-d-asp otbu-oh, r-2-9h-fluoren-9-yl methoxy carbonyl amino-4-tert-butoxy-4-oxobutanoic acid, fmoc-d-asp obut-oh, fmoc-d-aspartic acid beta-tert-butyl ester, n-alpha-fmoc-d-aspartic acid beta-t-butyl ester, 2r-4-tert-butoxy-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-oxobutanoic acid, fmoc-dasp otbu, ambotzfaa1309, fmoc-dasp otbu-oh, pubchem10009 | |
CC(C)(C)OC(=O)CC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O |
Specifications
112883-39-3 | |
MFCD00077050 | |
FODJWPHPWBKDON-UHFFFAOYNA-N | |
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-[(2-methylpropan-2-yl)oxy]-4-oxobutanoic acid | |
7018816 | |
98% |
C23H25NO6 | |
fmoc-d-asp otbu-oh, r-2-9h-fluoren-9-yl methoxy carbonyl amino-4-tert-butoxy-4-oxobutanoic acid, fmoc-d-asp obut-oh, fmoc-d-aspartic acid beta-tert-butyl ester, n-alpha-fmoc-d-aspartic acid beta-t-butyl ester, 2r-4-tert-butoxy-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-oxobutanoic acid, fmoc-dasp otbu, ambotzfaa1309, fmoc-dasp otbu-oh, pubchem10009 | |
CC(C)(C)OC(=O)CC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O | |
411.45 | |
411.45 | |
N-Fmoc-D-aspartic acid 4-tert-butyl ester |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only