missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-D-aspartic acid 4-tert-butyl ester, 98%, Thermo Scientific Chemicals

Product Code. 15487316
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112883-39-3
Molecular Formula C23H25NO6
Molecular Weight (g/mol) 411.45
MDL Number MFCD00077050
InChI Key FODJWPHPWBKDON-UHFFFAOYNA-N
Synonym fmoc-d-asp otbu-oh, r-2-9h-fluoren-9-yl methoxy carbonyl amino-4-tert-butoxy-4-oxobutanoic acid, fmoc-d-asp obut-oh, fmoc-d-aspartic acid beta-tert-butyl ester, n-alpha-fmoc-d-aspartic acid beta-t-butyl ester, 2r-4-tert-butoxy-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-oxobutanoic acid, fmoc-dasp otbu, ambotzfaa1309, fmoc-dasp otbu-oh, pubchem10009
PubChem CID 7018816
SMILES CC(C)(C)OC(=O)CC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Quantity 250 mg
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-[(2-methylpropan-2-yl)oxy]-4-oxobutanoic acid
Formula Weight 411.45
Percent Purity 98%
Chemical Name or Material N-Fmoc-D-aspartic acid 4-tert-butyl ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'