missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Fmoc-4-iodo-D-phenylalanine, 98%, Thermo Scientific Chemicals
25.62€ - 298.00€
Chemical Identifiers
CAS | 205526-29-0 |
---|---|
Molecular Formula | C24H20INO4 |
Molecular Weight (g/mol) | 513.33 |
MDL Number | MFCD00672557 |
InChI Key | LXOXXTQKKRJNNB-UHFFFAOYNA-N |
Synonym | fmoc-d-phe 4-i-oh, fmoc-4-iodo-d-phenylalanine, fmoc-d-4-iodophenylalanine, fmoc-d-4-iodophe, fmoc-4'-iodo-d-phe, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-iodophenyl propanoic acid, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-4-iodophenyl propanoic acid, ambotzfaa1584, fmoc-4-iodo-d-phe-oh, fmoc-d-phe 4-i-oh hplc |
PubChem CID | 16213204 |
SMILES | OC(=O)C(CC1=CC=C(I)C=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15479918
|
Thermo Scientific Alfa Aesar
H63292.MD |
250 mg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
15459918
|
Thermo Scientific Alfa Aesar
H63292.03 |
1 g |
74.80€
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15469918
|
Thermo Scientific Alfa Aesar
H63292.06 |
5 g |
298.00€
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
N-Fmoc-4-iodo-D-phenylalanine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-Fmoc-4-iodo-D-phenylalanine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Sparingly Soluble in water (6.7E-4 g/L) (25°C).
Notes
Light sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Chemical Identifiers
205526-29-0 | |
513.33 | |
LXOXXTQKKRJNNB-UHFFFAOYNA-N | |
16213204 |
C24H20INO4 | |
MFCD00672557 | |
fmoc-d-phe 4-i-oh, fmoc-4-iodo-d-phenylalanine, fmoc-d-4-iodophenylalanine, fmoc-d-4-iodophe, fmoc-4'-iodo-d-phe, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-iodophenyl propanoic acid, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-4-iodophenyl propanoic acid, ambotzfaa1584, fmoc-4-iodo-d-phe-oh, fmoc-d-phe 4-i-oh hplc | |
OC(=O)C(CC1=CC=C(I)C=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 |
Specifications
205526-29-0 | |
MFCD00672557 | |
fmoc-d-phe 4-i-oh, fmoc-4-iodo-d-phenylalanine, fmoc-d-4-iodophenylalanine, fmoc-d-4-iodophe, fmoc-4'-iodo-d-phe, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-iodophenyl propanoic acid, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-4-iodophenyl propanoic acid, ambotzfaa1584, fmoc-4-iodo-d-phe-oh, fmoc-d-phe 4-i-oh hplc | |
LXOXXTQKKRJNNB-UHFFFAOYNA-N | |
2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(4-iodophenyl)propanoic acid | |
16213204 | |
98% | |
Solid |
C24H20INO4 | |
9159551 | |
Sparingly Soluble in water (6.7E-4g/L) (25°C). | |
OC(=O)C(CC1=CC=C(I)C=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 | |
513.33 | |
513.32 | |
Light sensitive | |
N-Fmoc-4-iodo-D-phenylalanine |
Safety and Handling
missing translation for 'tsca' : No
missing translation for 'storageNote' : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only