missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-4-iodo-D-phenylalanine, 98%, Thermo Scientific Chemicals

Product Code. 15479918
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

N-Fmoc-4-iodo-D-phenylalanine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Fmoc-4-iodo-D-phenylalanine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Sparingly Soluble in water (6.7E-4 g/L) (25°C).

Notes
Light sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 205526-29-0
Molecular Formula C24H20INO4
Molecular Weight (g/mol) 513.33
MDL Number MFCD00672557
InChI Key LXOXXTQKKRJNNB-UHFFFAOYNA-N
Synonym fmoc-d-phe 4-i-oh, fmoc-4-iodo-d-phenylalanine, fmoc-d-4-iodophenylalanine, fmoc-d-4-iodophe, fmoc-4'-iodo-d-phe, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-iodophenyl propanoic acid, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-4-iodophenyl propanoic acid, ambotzfaa1584, fmoc-4-iodo-d-phe-oh, fmoc-d-phe 4-i-oh hplc
PubChem CID 16213204
SMILES OC(=O)C(CC1=CC=C(I)C=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Beilstein 9159551
Solubility Information Sparingly Soluble in water (6.7E-4g/L) (25°C).
IUPAC Name 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(4-iodophenyl)propanoic acid
Formula Weight 513.32
Percent Purity 98%
Quantity 250 mg
Sensitivity Light sensitive
Physical Form Solid
Chemical Name or Material N-Fmoc-4-iodo-D-phenylalanine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'