Learn More
N-Boc-sarcosine methyl ester, 97%, Thermo Scientific Chemicals
31.64€ - 446.00€
Chemical Identifiers
CAS | 42492-57-9 |
---|---|
Molecular Formula | C9H17NO4 |
Molecular Weight (g/mol) | 203.24 |
MDL Number | MFCD04973115 |
InChI Key | LTBDTYJYKRLTRN-UHFFFAOYSA-N |
Synonym | methyl 2-tert-butoxycarbonyl methyl amino acetate, n-boc-sarcosine methyl ester, boc-gly-ome, boc-sarcosine methyl ester, ltbdtyjykrltrn-uhfffaoysa, n-t-butoxycarbonyl-sarcosine methyl ester, n-tert-butoxycarbonylsarcosine methyl ester, methyl n-tert-butoxycarbonyl-n-methylglycinate, methyl 2-tert-butoxy carbonyl methyl amino acetate |
PubChem CID | 14018930 |
SMILES | COC(=O)CN(C)C(=O)OC(C)(C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15488838
|
Thermo Scientific Alfa Aesar
H62304.MD |
250 mg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
15468838
|
Thermo Scientific Alfa Aesar
H62304.03 |
1 g |
103.00€
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15478838
|
Thermo Scientific Alfa Aesar
H62304.06 |
5 g |
446.00€
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
N-Boc-sarcosine methyl ester is involved in the preparation of N-[(R, R)-(E)-1-arylmethyl-3-(2-oxo-azepan-3-yl) carbamoyl] allyl-N-methyl-3, 5-bis(trifluoromethyl) benzamides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-Boc-sarcosine methyl ester is involved in the preparation of N-[(R, R)-(E)-1-arylmethyl-3-(2-oxo-azepan-3-yl) carbamoyl] allyl-N-methyl-3, 5-bis(trifluoromethyl) benzamides.
Notes
Incompatible with oxidizing agents. Heat sensitive.
Chemical Identifiers
42492-57-9 | |
203.24 | |
LTBDTYJYKRLTRN-UHFFFAOYSA-N | |
14018930 |
C9H17NO4 | |
MFCD04973115 | |
methyl 2-tert-butoxycarbonyl methyl amino acetate, n-boc-sarcosine methyl ester, boc-gly-ome, boc-sarcosine methyl ester, ltbdtyjykrltrn-uhfffaoysa, n-t-butoxycarbonyl-sarcosine methyl ester, n-tert-butoxycarbonylsarcosine methyl ester, methyl n-tert-butoxycarbonyl-n-methylglycinate, methyl 2-tert-butoxy carbonyl methyl amino acetate | |
COC(=O)CN(C)C(=O)OC(C)(C)C |
Specifications
42492-57-9 | |
C9H17NO4 | |
MFCD04973115 | |
LTBDTYJYKRLTRN-UHFFFAOYSA-N | |
methyl 2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]acetate | |
14018930 | |
97% | |
N-Boc-sarcosine methyl ester |
Yellow | |
(CH3)3COCON(CH3)CH2CO2CH3 | |
methyl 2-tert-butoxycarbonyl methyl amino acetate, n-boc-sarcosine methyl ester, boc-gly-ome, boc-sarcosine methyl ester, ltbdtyjykrltrn-uhfffaoysa, n-t-butoxycarbonyl-sarcosine methyl ester, n-tert-butoxycarbonylsarcosine methyl ester, methyl n-tert-butoxycarbonyl-n-methylglycinate, methyl 2-tert-butoxy carbonyl methyl amino acetate | |
COC(=O)CN(C)C(=O)OC(C)(C)C | |
203.24 | |
203.24 | |
Liquid |
Safety and Handling
missing translation for 'tsca' : No
missing translation for 'storageNote' : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only