Learn More
N-Boc-sarcosine methyl ester, 97%, Thermo Scientific Chemicals
Artikelnummer. 15488838
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mg
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
N-Boc-sarcosine methyl ester is involved in the preparation of N-[(R, R)-(E)-1-arylmethyl-3-(2-oxo-azepan-3-yl) carbamoyl] allyl-N-methyl-3, 5-bis(trifluoromethyl) benzamides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Boc-sarcosine methyl ester is involved in the preparation of N-[(R, R)-(E)-1-arylmethyl-3-(2-oxo-azepan-3-yl) carbamoyl] allyl-N-methyl-3, 5-bis(trifluoromethyl) benzamides.
Notes
Incompatible with oxidizing agents. Heat sensitive.
TRUSTED_SUSTAINABILITY
Chemische Identifikatoren
CAS 42492-57-9
Molecular Formula C9H17NO4
Molecular Weight (g/mol) 203.24
MDL Number MFCD04973115
InChI Key LTBDTYJYKRLTRN-UHFFFAOYSA-N
Synonym methyl 2-tert-butoxycarbonyl methyl amino acetate, n-boc-sarcosine methyl ester, boc-gly-ome, boc-sarcosine methyl ester, ltbdtyjykrltrn-uhfffaoysa, n-t-butoxycarbonyl-sarcosine methyl ester, n-tert-butoxycarbonylsarcosine methyl ester, methyl n-tert-butoxycarbonyl-n-methylglycinate, methyl 2-tert-butoxy carbonyl methyl amino acetate
PubChem CID 14018930
SMILES COC(=O)CN(C)C(=O)OC(C)(C)C
Spezifikation
Color Yellow
Linear Formula (CH3)3COCON(CH3)CH2CO2CH3
IUPAC Name methyl 2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]acetate
Formula Weight 203.24
Percent Purity 97%
Quantity 250 mg
Physical Form Liquid
Chemical Name or Material N-Boc-sarcosine methyl ester