missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-L-glutamic acid, 98%, Thermo Scientific Chemicals
23.00€ - 37.60€
Chemical Identifiers
CAS | 2419-94-5 |
---|---|
Molecular Formula | C10H15NO6 |
Molecular Weight (g/mol) | 245.23 |
MDL Number | MFCD00037297 |
InChI Key | AQTUACKQXJNHFQ-LURJTMIESA-L |
Synonym | boc-glu-oh, boc-l-glutamic acid, n-boc-l-glutamic acid, boc-l-glu-oh, n-tert-butoxycarbonyl-l-glutamic acid, n-t-butoxycarbonyl-l-glutamic acid, n-alpha-t-boc-l-glutamic acid, n-tert-butoxycarbonyl glutamic acid, l-glutamic acid, n-1,1-dimethylethoxy carbonyl, n-alpha-t-butyloxycarbonyl-l-glutamic acid |
PubChem CID | 2724758 |
SMILES | CC(C)(C)OC(=O)N[C@@H](CCC([O-])=O)C([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15426358
|
Thermo Scientific Alfa Aesar
H59907.03 |
1 g |
23.00€
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15436358
|
Thermo Scientific Alfa Aesar
H59907.06 |
5 g |
37.60€
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2419-94-5 | |
245.23 | |
AQTUACKQXJNHFQ-LURJTMIESA-L | |
2724758 |
C10H15NO6 | |
MFCD00037297 | |
boc-glu-oh, boc-l-glutamic acid, n-boc-l-glutamic acid, boc-l-glu-oh, n-tert-butoxycarbonyl-l-glutamic acid, n-t-butoxycarbonyl-l-glutamic acid, n-alpha-t-boc-l-glutamic acid, n-tert-butoxycarbonyl glutamic acid, l-glutamic acid, n-1,1-dimethylethoxy carbonyl, n-alpha-t-butyloxycarbonyl-l-glutamic acid | |
CC(C)(C)OC(=O)N[C@@H](CCC([O-])=O)C([O-])=O |
Specifications
2419-94-5 | |
C10H15NO6 | |
2418563 | |
AQTUACKQXJNHFQ-LURJTMIESA-L | |
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanedioic acid | |
2724758 | |
98% |
∽110°C (decomposition) | |
MFCD00037297 | |
boc-glu-oh, boc-l-glutamic acid, n-boc-l-glutamic acid, boc-l-glu-oh, n-tert-butoxycarbonyl-l-glutamic acid, n-t-butoxycarbonyl-l-glutamic acid, n-alpha-t-boc-l-glutamic acid, n-tert-butoxycarbonyl glutamic acid, l-glutamic acid, n-1,1-dimethylethoxy carbonyl, n-alpha-t-butyloxycarbonyl-l-glutamic acid | |
CC(C)(C)OC(=O)N[C@@H](CCC([O-])=O)C([O-])=O | |
245.23 | |
247.25 | |
N-Boc-L-glutamic acid |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only