missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-glutamic acid, 98%, Thermo Scientific Chemicals

Product Code. 15436358
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2419-94-5
Molecular Formula C10H15NO6
Molecular Weight (g/mol) 245.23
MDL Number MFCD00037297
InChI Key AQTUACKQXJNHFQ-LURJTMIESA-L
Synonym boc-glu-oh, boc-l-glutamic acid, n-boc-l-glutamic acid, boc-l-glu-oh, n-tert-butoxycarbonyl-l-glutamic acid, n-t-butoxycarbonyl-l-glutamic acid, n-alpha-t-boc-l-glutamic acid, n-tert-butoxycarbonyl glutamic acid, l-glutamic acid, n-1,1-dimethylethoxy carbonyl, n-alpha-t-butyloxycarbonyl-l-glutamic acid
PubChem CID 2724758
SMILES CC(C)(C)OC(=O)N[C@@H](CCC([O-])=O)C([O-])=O

Specifications

Melting Point ∽110°C (decomposition)
Quantity 5 g
Beilstein 2418563
IUPAC Name (2S)-2-{[(tert-butoxy)carbonyl]amino}pentanedioate
Formula Weight 247.25
Percent Purity 98%
Chemical Name or Material N-Boc-L-glutamic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'