Learn More
N-Boc-4-cyano-D-phenylalanine, 98%, Thermo Scientific Chemicals
44.20€ - 659.00€
Chemical Identifiers
CAS | 146727-62-0 |
---|---|
Molecular Formula | C15H17N2O4 |
Molecular Weight (g/mol) | 289.31 |
MDL Number | MFCD00672527 |
InChI Key | RMBLTLXJGNILPG-GFCCVEGCSA-M |
Synonym | boc-d-phe 4-cn-oh, r-n-boc-4-cyanophenylalanine, boc-4-cyano-d-phenylalanine, boc-d-4-cyanophenylalanine, r-2-tert-butoxycarbonyl amino-3-4-cyanophenyl propanoic acid, 2r-2-tert-butoxycarbonyl amino-3-4-cyanophenyl propanoic acid, d-phenylalanine, 4-cyano-n-1,1-dimethylethoxy carbonyl, boc-d-4-cyanophe, boc-4-cyano-d-phe-oh, boc-d-4-cn-phe-oh |
PubChem CID | 2734494 |
IUPAC Name | (2R)-3-(4-cyanophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid |
SMILES | CC(C)(C)OC(=O)N[C@H](CC1=CC=C(C=C1)C#N)C([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15419398
|
Thermo Scientific Alfa Aesar
H62947.MD |
250 mg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
15499388
|
Thermo Scientific Alfa Aesar
H62947.03 |
1 g |
160.00€
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15409398
|
Thermo Scientific Alfa Aesar
H62947.06 |
5 g |
659.00€
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
N-Boc-4-cyano-D-phenylalanine is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-Boc-4-cyano-D-phenylalanine is used as pharmaceutical intermediate.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Chemical Identifiers
146727-62-0 | |
289.31 | |
RMBLTLXJGNILPG-GFCCVEGCSA-M | |
2734494 | |
CC(C)(C)OC(=O)N[C@H](CC1=CC=C(C=C1)C#N)C([O-])=O |
C15H17N2O4 | |
MFCD00672527 | |
boc-d-phe 4-cn-oh, r-n-boc-4-cyanophenylalanine, boc-4-cyano-d-phenylalanine, boc-d-4-cyanophenylalanine, r-2-tert-butoxycarbonyl amino-3-4-cyanophenyl propanoic acid, 2r-2-tert-butoxycarbonyl amino-3-4-cyanophenyl propanoic acid, d-phenylalanine, 4-cyano-n-1,1-dimethylethoxy carbonyl, boc-d-4-cyanophe, boc-4-cyano-d-phe-oh, boc-d-4-cn-phe-oh | |
(2R)-3-(4-cyanophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid |
Specifications
146727-62-0 | |
MFCD00672527 | |
9339089 | |
RMBLTLXJGNILPG-GFCCVEGCSA-M | |
(2R)-3-(4-cyanophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid | |
2734494 | |
98% |
C15H17N2O4 | |
UN3439 | |
boc-d-phe 4-cn-oh, r-n-boc-4-cyanophenylalanine, boc-4-cyano-d-phenylalanine, boc-d-4-cyanophenylalanine, r-2-tert-butoxycarbonyl amino-3-4-cyanophenyl propanoic acid, 2r-2-tert-butoxycarbonyl amino-3-4-cyanophenyl propanoic acid, d-phenylalanine, 4-cyano-n-1,1-dimethylethoxy carbonyl, boc-d-4-cyanophe, boc-4-cyano-d-phe-oh, boc-d-4-cn-phe-oh | |
CC(C)(C)OC(=O)N[C@H](CC1=CC=C(C=C1)C#N)C([O-])=O | |
289.31 | |
290.31 | |
N-Boc-4-cyano-D-phenylalanine |
Safety and Handling
GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only