missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-4-cyano-D-phenylalanine, 98%, Thermo Scientific Chemicals

Product Code. 15409398
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

N-Boc-4-cyano-D-phenylalanine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-4-cyano-D-phenylalanine is used as pharmaceutical intermediate.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 146727-62-0
Molecular Formula C15H17N2O4
Molecular Weight (g/mol) 289.31
MDL Number MFCD00672527
InChI Key RMBLTLXJGNILPG-GFCCVEGCSA-M
Synonym boc-d-phe 4-cn-oh, r-n-boc-4-cyanophenylalanine, boc-4-cyano-d-phenylalanine, boc-d-4-cyanophenylalanine, r-2-tert-butoxycarbonyl amino-3-4-cyanophenyl propanoic acid, 2r-2-tert-butoxycarbonyl amino-3-4-cyanophenyl propanoic acid, d-phenylalanine, 4-cyano-n-1,1-dimethylethoxy carbonyl, boc-d-4-cyanophe, boc-4-cyano-d-phe-oh, boc-d-4-cn-phe-oh
PubChem CID 2734494
IUPAC Name (2R)-3-(4-cyanophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)N[C@H](CC1=CC=C(C=C1)C#N)C([O-])=O

Specifications

Quantity 5 g
UN Number UN3439
Beilstein 9339089
Formula Weight 290.31
Percent Purity 98%
Chemical Name or Material N-Boc-4-cyano-D-phenylalanine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'