missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-3,4-dichloro-L-phenylalanine, 95%, Thermo Scientific Chemicals
49.20€ - 482.00€
Chemical Identifiers
CAS | 80741-39-5 |
---|---|
Molecular Formula | C14H16Cl2NO4 |
Molecular Weight (g/mol) | 333.19 |
MDL Number | MFCD00273436 |
InChI Key | UGZIQCCPEDCGGN-NSHDSACASA-M |
Synonym | boc-3,4-dichloro-l-phenylalanine, boc-phe 3,4-dicl-oh, boc-l-3,4-dichlorophe, s-2-tert-butoxycarbonyl amino-3-3,4-dichlorophenyl propanoic acid, boc-l-3,4-dichlorophenylalanine, 2s-2-tert-butoxycarbonyl amino-3-3,4-dichlorophenyl propanoic acid, 2s-2-tert-butoxycarbonylamino-3-3,4-dichlorophenyl propanoic acid, boc-3,4-dichloro-phe-oh, boc-phe 3,4-cl2-oh, boc-l-3,4-dichloro phenylalanine |
PubChem CID | 2734501 |
IUPAC Name | (2S)-3-(3,4-dichlorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid |
SMILES | CC(C)(C)OC(=O)N[C@@H](CC1=CC=C(Cl)C(Cl)=C1)C([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15458547
|
Thermo Scientific Alfa Aesar
H52053.MD |
250 mg |
49.20€
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15438547
|
Thermo Scientific Alfa Aesar
H52053.03 |
1 g |
163.00€
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15448547
|
Thermo Scientific Alfa Aesar
H52053.06 |
5 g |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
80741-39-5 | |
333.19 | |
UGZIQCCPEDCGGN-NSHDSACASA-M | |
2734501 | |
CC(C)(C)OC(=O)N[C@@H](CC1=CC=C(Cl)C(Cl)=C1)C([O-])=O |
C14H16Cl2NO4 | |
MFCD00273436 | |
boc-3,4-dichloro-l-phenylalanine, boc-phe 3,4-dicl-oh, boc-l-3,4-dichlorophe, s-2-tert-butoxycarbonyl amino-3-3,4-dichlorophenyl propanoic acid, boc-l-3,4-dichlorophenylalanine, 2s-2-tert-butoxycarbonyl amino-3-3,4-dichlorophenyl propanoic acid, 2s-2-tert-butoxycarbonylamino-3-3,4-dichlorophenyl propanoic acid, boc-3,4-dichloro-phe-oh, boc-phe 3,4-cl2-oh, boc-l-3,4-dichloro phenylalanine | |
(2S)-3-(3,4-dichlorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid |
Specifications
80741-39-5 | |
MFCD00273436 | |
boc-3,4-dichloro-l-phenylalanine, boc-phe 3,4-dicl-oh, boc-l-3,4-dichlorophe, s-2-tert-butoxycarbonyl amino-3-3,4-dichlorophenyl propanoic acid, boc-l-3,4-dichlorophenylalanine, 2s-2-tert-butoxycarbonyl amino-3-3,4-dichlorophenyl propanoic acid, 2s-2-tert-butoxycarbonylamino-3-3,4-dichlorophenyl propanoic acid, boc-3,4-dichloro-phe-oh, boc-phe 3,4-cl2-oh, boc-l-3,4-dichloro phenylalanine | |
CC(C)(C)OC(=O)N[C@@H](CC1=CC=C(Cl)C(Cl)=C1)C([O-])=O | |
333.19 | |
334.2 | |
N-Boc-3,4-dichloro-L-phenylalanine |
C14H16Cl2NO4 | |
6426101 | |
UGZIQCCPEDCGGN-NSHDSACASA-M | |
(2S)-3-(3,4-dichlorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid | |
2734501 | |
95% |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only