missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-3,4-dichloro-L-phenylalanine, 95%, Thermo Scientific Chemicals

Product Code. 15448547
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80741-39-5
Molecular Formula C14H16Cl2NO4
Molecular Weight (g/mol) 333.19
MDL Number MFCD00273436
InChI Key UGZIQCCPEDCGGN-NSHDSACASA-M
Synonym boc-3,4-dichloro-l-phenylalanine, boc-phe 3,4-dicl-oh, boc-l-3,4-dichlorophe, s-2-tert-butoxycarbonyl amino-3-3,4-dichlorophenyl propanoic acid, boc-l-3,4-dichlorophenylalanine, 2s-2-tert-butoxycarbonyl amino-3-3,4-dichlorophenyl propanoic acid, 2s-2-tert-butoxycarbonylamino-3-3,4-dichlorophenyl propanoic acid, boc-3,4-dichloro-phe-oh, boc-phe 3,4-cl2-oh, boc-l-3,4-dichloro phenylalanine
PubChem CID 2734501
IUPAC Name (2S)-3-(3,4-dichlorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)N[C@@H](CC1=CC=C(Cl)C(Cl)=C1)C([O-])=O

Specifications

Quantity 5 g
Beilstein 6426101
Formula Weight 334.2
Percent Purity 95%
Chemical Name or Material N-Boc-3,4-dichloro-L-phenylalanine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'