Learn More
Mecmesin™ Multitest-i Computer Controlled Force Test Stand
Suitable for production testing.
1582.50€ - 11925.00€
Specifications
Product Type Test Stand
For Use With (Application) Suitable for production testing
View More Specs
Products 2
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
11953998
Encompass
View Documents
Mecmesin™
879003
Force: 50N
1582.50€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 27-01-2025
to see stock.
Add to basket
11933908
Encompass
View Documents
Mecmesin™
805102
Force: 2.5N
11925.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 27-01-2025
to see stock.
Add to basket
Description
Description
  • Build Program
  • Design and tailor your force or tensile test
  • Intuitive, easy-to-learn user interface
  • Toolbars simplify test management
  • Create sample pass/fail criteria
  • Automatically detects parameters e.g. peak load, average load, etc
  • Contains a library of typical test procedures
  • Displacement
  • Break or time
Specifications
Specifications
Test Stand
Suitable for production testing