Learn More
Mecmesin™ Multitest-i Computer Controlled Force Test Stand
Suitable for production testing.
Brand: Mecmesin™ 805102
This item is not returnable.
View return policy
Description
- Build Program
- Design and tailor your force or tensile test
- Intuitive, easy-to-learn user interface
- Toolbars simplify test management
- Create sample pass/fail criteria
- Automatically detects parameters e.g. peak load, average load, etc
- Contains a library of typical test procedures
- Displacement
- Break or time
Specifications
Test Stand | |
Force: 2.5N | |
Suitable for production testing |