Learn More
Mecmesin™ Multifunction Force/Torque Indicator
Features high speed sampling rate.
Brand:  Mecmesin™ 851401
Encompass
Product Code. 11947979
  • POA / Each
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Récupel-bijdrage uitgesloten
Description
Description
  • Advanced indicator with precision external force, torque sensors
  • High-speed 5000Hz sampling rate
  • Audio/visual alarms
  • RS-232 interface
  • Graphical display load-bar graph
  • Live and peak readings simultaneously
  • Takes high-accuracy measurements with precision external smart transducers
  • Meter automatically recognizes transducers—no calibration or setup
  • Large, backlit display provides graphical representation
  • Sampling rate: 5000 HZ
  • Data storage: 500
  • Output: RS-232
Specifications
Specifications
Indicator
Multifunction force/torque indicator
Takes high-accuracy measurements with precision external smart transducer
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Mecmesin™ Multifunction Force/Torque Indicator