Learn More
Chemetrics™ Multi-Analyte Photometer V–2000 for Water Quality Analysis
Advanced portable microprocessor-based LED colorimeter
Brand:  Chemetrics™ V-2000
Product Code. 11946208
  • 1560.00€ / Each
Estimated Shipment: 23-05-2025
to see stock.
Explore more special offers
Add to basket
Récupel-bijdrage uitgesloten
Description
Description
Portable water analyzer automatically tests pre-programmed (50+) analytes using CHEMetrics Vacu-vials™ self-filling, pre-measured ampoules. Operator safety is ensured with no sample preparation, mixing or chemical contact. As new tests are available, a simple upload procedure from the CHEMetrics website updates this analyzer with the most recent programs. Uploading takes a few minutes and keeps the V-2000 constantly current. Up to ten user-created custom methods can be stored in the photometer's memory. The simple interface guides a user through setup and measurement.
  • Field Portable
  • Battery operated
  • Lightweight
  • Rugged
  • Waterproof
Specifications
Specifications
Photometer
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Chemetrics™ Multi-Analyte Photometer V–2000 for Water Quality Analysis > Wavelength: 420, 520, 580, 610nm