missing translation for 'onlineSavingsMsg'
Learn More
MRS 2179 tetrasodium salt, Tocris Bioscience™ Product Code.: 18064382

MRS 2179 tetrasodium salt, Tocris Bioscience™

Product Code. 18064382
50 mg
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Product Code. 18064382

Brand: Tocris Bioscience 0900/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective P2Y1 antagonist

A competitive antagonist at P2Y1 receptors (KB = 100 nM). Selective over P2X1 (IC50 = 1.15 μM), P2X3 (IC50 = 12.9 μM), P2X2, P2X4, P2Y2, P2Y4 and P2Y6 receptors. Inhibits the upregulation of NTPDase1 by ATPγS.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1454889-37-2
Molecular Formula C11H17N5Na4O9P2
Molecular Weight (g/mol) 517.19
InChI Key HLJPLHFSEAJJGC-ZKRIHRHSSA-N
Synonym 2'-Deoxy-N6-methyladenosine 3', 5'-bisphosphate tetrasodium salt
PubChem CID 131848299
IUPAC Name [(2R,3S,5R)-5-[6-(methylamino)purin-9-yl]-2-(phosphonooxymethyl)oxolan-3-yl] dihydrogen phosphate;sodium
SMILES CNC1=NC=NC2=C1N=CN2C3CC(C(O3)COP(=O)(O)O)OP(=O)(O)O.[Na].[Na].[Na].[Na]

Specifications

Quantity 50 mg
Formula Weight 513.16
Percent Purity >98%
Chemical Name or Material MRS 2179 tetrasodium salt
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
MRS 2179 tetrasodium salt, Tocris Bioscience™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.