missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Goat Serum: Normal, Sterile
Suitable replacement for FBS on most cell lines and primary cultures for Biomedical Research
107.06€
Specifications
Certifications/Compliance | U.S. regulations |
---|---|
Color | Red |
Content And Storage | -20°C |
Species | Goat |
Product Type | Goat Serum |
Description
Specifications
U.S. regulations | |
-20°C | |
Goat Serum | |
0.2 μm filtration | |
<20mg/dL | |
6.5 to 8 |
Red | |
Goat | |
Sterile | |
Immunohistochemistry | |
No |