missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Goat Serum: Normal, Sterile

Suitable replacement for FBS on most cell lines and primary cultures for Biomedical Research

Brand:  MP Biomedicals™ 0219135680

86.79 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 11450422

  • 107.06€ / 100mL
Estimated Shipment: 09-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

TRUSTED_SUSTAINABILITY
Specifications

Specifications

U.S. regulations
-20°C
Goat Serum
Sterile
Immunohistochemistry
No
Red
Goat
100 mL
0.2 μm filtration
<20mg/dL
6.5 to 8
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions