Learn More
MP Biomedicals™ Biotrans(+) Nylon Membranes, 30cmx3m Roll, 0.45um
Used for alkaline, Western, and Improved Southern Blots. Reinforced nylon 66 membranes for use in DNA, RNA and protein transfers. The physical characteristics of BIOTRANS membranes make them especially useful as a transfer medium.
Brand:  MP Biomedicals™ 01810205
Product Code. 11343489
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Specifications
30 cm
3 m
Roll
0.45 μm