missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse IFN-beta DuoSet ELISA
Mouse IFN-beta DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Brand: R&D Systems™ DY8234-05
Includes: Capture antibody, detection antibody, recombinant standard, streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP) and development protocols
Description
The Mouse IFN-beta DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies mouse IFN-beta.
Specifications
ELISA Kit | |
IFN-beta | |
ELISA | |
Cell Culture Supernatant, Serum, Plasma | |
Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
Mouse | |
96-well Plates | |
3456 | |
Fibroblast interferon, IFB, IFBIFNB, IFF, IFNB, IFNB1, IFNbeta, IFN-beta, interferon beta, interferon, beta 1, fibroblast, MGC96956 |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
R&D Systems™ Mouse IFN-beta DuoSet ELISA > Mouse IFN-beta DuoSet ELISA Kit
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction