Learn More
Invitrogen™ Glutathione Ethyl Ester, Biotin Amide (BioGEE) (Glutathiolation Detection Reagent) - Special Packaging
Glutathiolation detection reagent
Brand:  Invitrogen™ G36000
335.16 EUR valid until 2025-03-28
Use promo code "24107" to get your promotional price.
Product Code. 11534866
  • 446.00€ / 1000µg
Explore more special offers
Description
Description
Biotinylated glutathione ethyl ester (BioGEE) is cell-permeant, biotinylated glutathione analog for detection of glutathiolation Under conditions of oxidative stress, cells may transiently incorporate glutathione into proteins. Stressed cells incubated in BioGEE will also incorporate this biotinylated glutathione derivative into proteins, facilitating the identification of oxidation-sensitive proteins.
  • Glutathiolation levels can be detecte with fluorescent streptavidin conjugate using either flow cytometry or fluorescence microscopy
  • Proteins glutathiolated with BioGEE can also be extracted and analyzed by mass spectrometry or by Western blotting methods in conjunction with fluorophore- or enzyme-labeled streptavidin conjugates
Cell Analysis, Cell Metabolism, Cell Viability, Proliferation and Function, Glutathione Detection, Nitro-Oxidative Stress, Reactive Oxygen Species
Order Info
Shipping Condition: Room Temperature
TRUSTED_SUSTAINABILITY
Specifications
Specifications
10 x 100 μg
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2810931 Certificate TypeCertificate of Analysis Date13/07/2024 Catalog NumberG36000
Lot Number2595618 Certificate TypeCertificate of Analysis Date14/04/2023 Catalog NumberG36000
Lot Number2413438 Certificate TypeCertificate of Analysis Date01/12/2021 Catalog NumberG36000
Lot Number2311924 Certificate TypeCertificate of Analysis Date18/01/2021 Catalog NumberG36000
Lot Number2208540 Certificate TypeCertificate of Analysis Date17/10/2020 Catalog NumberG36000
Product Title
Invitrogen™ Glutathione Ethyl Ester, Biotin Amide (BioGEE) (Glutathiolation Detection Reagent) - Special Packaging > 10 x 100ug