Learn More
3M™ Molecular Detection Assay 2
Used with the 3M™ Molecular Detection System for the rapid and specific detection of shiga toxin-producing e.coli species in enriched food and environmental samples.
Brand: 3M™ 7100194040
Description
- One assay protocol and fewer steps than most tests reduces repetitive tasks and makes it easy to train employees.
- Ready-to-use and pre-dispensed reagents provide convenience and minimize the chance of human error.
- Color-coded assay tubes by organism and matching software screens allow for better tracking and easy identification.
- The ability to run all assays simultaneously and the flexibility to test up to 96 samples in each run helps you operate more efficiently.
- Real-time amplification and detection process identifies positive results in as early as 15 minutes and allows you to make critical decisions faster.
Specifications
STEC Gene Screen | |
Microbiology Media | |
Food and Beverage, Environmental Test | |
48 Pack |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.