missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ ML 162
Inhibits glutathione peroxidase (GPX4); induces ferroptosis
185.74€ - 728.92€
Specifications
Chemical Name or Material | 2-[(Chloroacetyl)(3-chloro-4-methoxyphenyl)amino]-N-(2-phenylethyl)-2-thien-2-ylacetamide |
---|---|
CAS | 1035072-16-2 |
Target | Glutathione Peroxidase 4/GPX4 |
Molecular Formula | C23H22Cl2N2O3S |
Purity | 0.98 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30000030
|
Tocris Bioscience™
7821/10 |
10 mg |
185.74€
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30000010
|
Tocris Bioscience™
7821/50 |
50 mg |
728.92€
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ML 162 is a covalent glutathione peroxidase 4 (GPX4) inhibitor. It induces ferroptosis in vivo. It is selectively lethal to mutant RAS oncogene-expressing cell lines (IC50 = 25 and 578 nM for HRASG12V-expressing and wild-type BJ fibroblasts, respectively).Specifications
2-[(Chloroacetyl)(3-chloro-4-methoxyphenyl)amino]-N-(2-phenylethyl)-2-thien-2-ylacetamide | |
Glutathione Peroxidase 4/GPX4 | |
0.98 |
1035072-16-2 | |
C23H22Cl2N2O3S |
Safety and Handling
Recommended Storage : Store at -20°C