missing translation for 'onlineSavingsMsg'
Learn More
ML 145, Tocris Bioscience™ Product Code.: 18046032

ML 145, Tocris Bioscience™

Product Code. 18046032
10 mg
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Product Code. 18046032

Brand: Tocris Bioscience 4172/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

GPR35 antagonist

Selective antagonist for the GPR35 orphan receptor GPCR (IC50 = 20.1 nM). Over 1000-fold more selective for GPR35 compared to GPR55 antagonists.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1164500-72-4
Molecular Formula C24H22N2O5S2
Molecular Weight (g/mol) 482.569
InChI Key COFMYJWNXSFLKQ-QIROLCGISA-N
Synonym 2-hydroxy-4-4-5-2-methyl-3-phenyl-2-propen-1-ylidene-4-oxo-2-thioxo-1,3-thiazolidin-3-yl butanoyl amino benzoic acid, 2-hydroxy-4-4-5z-5-e-2-methyl-3-phenylprop-2-enylidene-4-oxo-2-sulfanylidene-1,3-thiazolidin-3-yl butanoylamino benzoic acid, 2-hydroxy-4-4-5z-5-2e-2-methyl-3-phenylprop-2-en-1-ylidene-4-oxo-2-thioxo-1,3-thiazolidin-3-yl butanoyl amino benzoic acid, 2-hydroxy-4-4-5z-5-e-2-methyl-3-phenylprop-2-enylidene-4-oxo-2-sulfanylidene-3-thiazolidinyl-1-oxobutyl amino benzoic acid, 2-hydroxy-4-4-5z-4-keto-5-e-2-methyl-3-phenyl-prop-2-enylidene-2-thioxo-thiazolidin-3-yl butanoylamino benzoic acid, 2-hydroxy-4-4-5z-5-2e-2-methyl-3-phenylprop-2-en-1-ylidene-4-oxo-2-sulfanylidene-1,3-thiazolidin-3-yl butanamido benzoic acid, 4-4-5z-5-e-2-methyl-3-phenyl-prop-2-enylidene-4-oxidanylidene-2-sulfanylidene-1,3-thiazolidin-3-yl butanoylamino-2-oxidanyl-benzoic acidShow MoreShow Less
PubChem CID 2286812
IUPAC Name 2-hydroxy-4-[4-[(5Z)-5-[(E)-2-methyl-3-phenylprop-2-enylidene]-4-oxo-2-sulfanylidene-1,3-thiazolidin-3-yl]butanoylamino]benzoic acid
SMILES CC(=CC1=CC=CC=C1)C=C2C(=O)N(C(=S)S2)CCCC(=O)NC3=CC(=C(C=C3)C(=O)O)O

Specifications

Quantity 10 mg
Formula Weight 482.57
Percent Purity >98%
Chemical Name or Material ML 145
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
ML 145, Tocris Bioscience™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.