Learn More
behr Labor-Technik™ Mixed-Bed Ion Exchanger Made Of Stainless Steel
Standard cartridge made of V4A stainless steel for general purpose application: To supply laboratory glassware washers, for general laboratory maintenance and for small industrial consumers. For subsequent demineralization of reverse osmosis systems.
Brand:  behr Labor-Technik™ B00223482
 View more versions of this product
Product Code. 10799351
  • 2595.00€ / Each
Estimated Shipment: 17-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Optimum exploitation of the ion exchanger capacity due to absolute uniform water distribution in the unit
  • Flow rates up to 700 l/h
  • The unique behropur™ jet nozzle system distributes the raw water over the entire resin bed. This ensures an optimum exchange capacity and quality
  • Can be connected to the raw water network directly and without a pressure reducer
  • Resistant to alternating pressure
  • Hard rubber collars vulcanized to the base and top of the unit offer effective protection
  • Convenient transport thanks to the handles embedded into the top part of the unit
  • Conductance is controlled directly on the unit (installed measuring equipment) or with remote location meter
  • Also available with shutoff when the limited value is reached and limit control
  • Specifications
    Specifications
    74 mm
    E40d Pressure resistant mixed-bed unit made of V 4 A special alloy, permanent pressure resistant to 10 bar, complete with conductivity meter
    500 L/hr.