Learn More
behr Labor-Technik™ Mixed-Bed Ion Exchanger Made Of Stainless Steel
Standard cartridge made of V4A stainless steel for general purpose application: To supply laboratory glassware washers, for general laboratory maintenance and for small industrial consumers. For subsequent demineralization of reverse osmosis systems.
2595.00€ - 4110.00€
Products 2
Product Code Brand Flow Rate Price Quantity & Availability  
Product Code Brand Flow Rate Price Quantity & Availability  
10799351
View Documents
behr Labor-Technik™
B00223482
500 L/hr.
2595.00€
Each
Estimated Shipment: 12-02-2025
to see stock.
Add to basket
10598092
View Documents
behr Labor-Technik™
B00223483
700 L/hr.
4110.00€
Each
Estimated Shipment: 03-01-2025
to see stock.
Add to basket
Description
Description
  • Optimum exploitation of the ion exchanger capacity due to absolute uniform water distribution in the unit
  • Flow rates up to 700 l/h
  • The unique behropur™ jet nozzle system distributes the raw water over the entire resin bed. This ensures an optimum exchange capacity and quality
  • Can be connected to the raw water network directly and without a pressure reducer
  • Resistant to alternating pressure
  • Hard rubber collars vulcanized to the base and top of the unit offer effective protection
  • Convenient transport thanks to the handles embedded into the top part of the unit
  • Conductance is controlled directly on the unit (installed measuring equipment) or with remote location meter
  • Also available with shutoff when the limited value is reached and limit control
  • Videos