Learn More
Macherey-Nagel™ Mix VI Diamino/C18 ec Clean-up Mix
Brand: Macherey-Nagel™ 730974
This item is not returnable.
View return policy
Specifications
Chromabond QuEChERS Clean-up Mixes (EN 15662) | |
Diamino/C18 ec Clean-up Mix | |
15 mL | |
50 Pack |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 7
Product Content Correction
Product Title
Macherey-Nagel™ Mix VI Diamino/C18 ec Clean-up Mix > Volume: 15mL