Learn More
Merck Minicon CS15
For the concentration of urine and cerebrospinal fluid for enrichment of protein analytes (e.g., Bence Jones proteins in urine). Minicon CS15 is a static concentrator, requiring no accessories.
Brand: Merck 9051
This item is not returnable.
View return policy
Description
- Low hold-up volume
- Concentration factor up to 100x
- Absorbent pulls solvent and salts through ultrafilter, concentrating the sample
Specifications
Clear | |
Concentration of Urine and Cerebrospinal Fluid for Enrichment of Protein Analytes (e.g., Bence Jones Proteins in Urine) | |
Minicon CS15 |
Minicon CS15 | |
50/Pack | |
2.5 mL |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Product Title
Merck Minicon CS15 > Volume: 2.5mL; For use with: Minicon CS15