Learn More
Mecmesin™ Miniature Button Load Cell
Used for compression only, and is ideal for applications in restricted spaces.
Brand:  Mecmesin™ 878013
Encompass
 View more versions of this product
Product Code. 11943978
  • POA / Each
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Récupel-bijdrage uitgesloten
Description
Description
  • Use the S type load cells where space is not limited; dedicated fixtures attach via threaded holes. Static torque screwdrivers may be handheld or mounted.
Specifications
Specifications
5000 N
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Mecmesin™ Miniature Button Load Cell