Learn More
Ansell™ Microchem™ 3000 Aprons with Sleeves Product Code.: 15506604
Add to basket
Ansell™ Microchem™ 3000 Aprons with Sleeves
Product Code. 15506604 Shop All Ansell Products
Medium
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Product Code. 15506604
Brand: Ansell™ YE30W0021403
Edge
Add to basket
Add to basket
Constructed of a multi-layer barrier laminate. Ansell Microchem™ 3000 Aprons with Sleeves with welded seams provide liquid tight connections.
  • Apron with Sleeves
  • Rear velcro fastening
  • Elasticated wrists
  • Welded seams
  • Multi-layer barrier fabric effective against numerous chemicals
  • Highly visible bright yellow for improved worker safety
  • Lightweight yet durable
  • Anti-static according to EN 1149-5
TRUSTED_SUSTAINABILITY
Specifications
Type Protective Apron
Size Medium
Material Multi-layer Barrier Laminate Fabric
Color Yellow
Closure Type Hook and Loop (Rear)
Cuff Style Elastic
No. of Pockets None
Certifications/Compliance Type PB 3/4 EN 14605, Type PB(3)-B, PB(4)-B EN 14126, EN1149-1
Seam Type Ultrasonically Welded
Product Content Correction
Product Title
Ansell™ Microchem™ 3000 Aprons with Sleeves >