missing translation for 'onlineSavingsMsg'
Learn More

SI Analytics™ Micro-Ubbelohde Viscometers

SI Analytics™ Micro-Ubbelohde Viscometers feature a suspended ball level for determination of absolute and relative kinematic viscosity of liquids with Newtonian flow behavior.

528.00€

Specifications

Product Type Ubbelohde Micro Viscometer
Calibrated Yes
Certifications/Compliance DIN 51562, Part 2
Material Glass
Length (Metric) 290 mm
View More Specs

Products 2
Product Code Brand Viscosity Range Price Quantity & Availability  
Product Code Brand Viscosity Range Price Quantity & Availability  
10632441
View accessory products View Documents
SI Analytics™
285401025
4 to 60 cSt
528.00€
Each
Estimated Shipment: 31-01-2025
to see stock.
Add to basket
 
10766911
View accessory products View Documents
SI Analytics™
285401041
40 to 800 cSt
528.00€
Each
Estimated Shipment: 06-02-2025
to see stock.
Add to basket
 
Description

Description

  • Calibrated, with constant, for manual measurement
  • Ideal for transparent liquids manual measurement, foaming liquids, liquid mixture with highly volatile components, minimum measurement substance and/or rinsing agent quantities, and high-temperature or low temperature measurements
  • Delivered with manufacturer's certificate in accordance with DIN 55 350, Part 18
  • All viscometers are provided with ring marks
  • Due to their design, these viscometers are especially suitable for measurement of small liquid quantities and for particularly short running times
  • This ensures that viscometers for automatic measurements can also be checked by means of manual measurements
  • This constant is determined by multiplication of the constant K with the correction factor F
Specifications

Specifications

Ubbelohde Micro Viscometer
DIN 51562, Part 2
290 mm
3 to 4 mL
Yes
Glass
Manual measurements
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.