Learn More
SI Analytics™ Micro-Ubbelohde Viscometers
SI Analytics™ Micro-Ubbelohde Viscometers feature a suspended ball level for determination of absolute and relative kinematic viscosity of liquids with Newtonian flow behavior.
Brand:  SI Analytics™ 285401025
 View more versions of this product
Product Code. 10632441
  • 528.00€ / Each
Explore more special offers
Description
Description
  • Calibrated, with constant, for manual measurement
  • Ideal for transparent liquids manual measurement, foaming liquids, liquid mixture with highly volatile components, minimum measurement substance and/or rinsing agent quantities, and high-temperature or low temperature measurements
  • Delivered with manufacturer's certificate in accordance with DIN 55 350, Part 18
  • All viscometers are provided with ring marks
  • Due to their design, these viscometers are especially suitable for measurement of small liquid quantities and for particularly short running times
  • This ensures that viscometers for automatic measurements can also be checked by means of manual measurements
  • This constant is determined by multiplication of the constant K with the correction factor F
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ubbelohde Micro Viscometer
Yes
0.1
Glass
Manual measurements
4 to 60 cSt
0.70 mm diameter
DIN 51562, Part 2
290 mm
3 to 4 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products