missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ Solvent Manager Set, for Mettler Toledo™ Titrators

For use with V20/C20 Karl Fischer titrators.

Brand:  Mettler Toledo™ 51105600

Product Code. 12858241

  • 1575.00€ / 1 set
Estimated Shipment: 28-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

For use with V20/C20 Karl Fischer titrators.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Solvent Manager Set for V20/C20 Karl Fischer Titrators
Titration
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Mettler Toledo™ Solvent Manager Set, for Mettler Toledo™ Titrators > Solvent Manager Set for V20/C20 Karl Fischer Titrators

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.